Learn More
Epredia™ ClearVue™ Coverslipper Coverslip Hoppers
Designed for use with the Epredia™ ClearVue™ coverlipper. ClearVue™ hoppers streamline coverslipping workflow.
Brand: Epredia™ A79210050
120.12 EUR valid until 2024-12-31
Use promo code "21628" to get your promotional price.
This item is not returnable.
View return policy
Description
- Makes integration into the laboratory effortless
- Ships preloaded with 500 or 575 slips each
- Inserts quickly into the Epredia™ ClearVue™ coverslipper without workflow interruption
- Requires no manual handling of glass
- Dispenses a clean coverslip each and every time
Specifications
Coverslip Hopper | |
2 x 500 Hoppers/Coverslips | |
0.16 to 0.19 mm (# 1.5) |
Epredia ClearVue Coverslipper | |
24 mm | |
40 mm |