Learn More
Liofilchem™ Clean Test
Product Code. 11984359
Encompass_Preferred
Click to view available options
Detects:
Protein Fat And Sugar Residues
Unit Size:
Pack of 50
Product Code. 11984359
Brand: Liofilchem™ 86001
Only null left
Add to basket
Add to basket
Rapid test for the hygiene control of work surfaces
  • Clean test is a swab for the detection of protein, fat and sugar residues suitable for the check of work surfaces cleaning level.
Specifications
Detects Protein Fat And Sugar Residues
Manufacturing Origin Italy
Product Type Clean Test
Sterility Non-sterile
For Use With (Application) For The Hygiene Control Of Work Surfaces