Learn More
Hirschmann™ Class A Brown Graduated USP Measuring Pipettes
missing translation for 'orderingAttributeHoverText'
Volume (Metric):
1 mL
2 mL
3 mL
8 mL
9 mL
25 mL
50 mL
100 mL
missing translation for 'unitSize'
Pack of 12
Pack of 6
Description
- With dated batch identification
- Manufactured from AR glass™ soda-lime glass with a smooth, non-porous surface
- With volume mark
- Brown graduation
Specifications
Specifications
| Class | A |
| Material | AR-GLAS |
| For Use With (Application) | Volumetric accuracy in pipetting |
| Graduated | Yes |
| Graduation Color | Brown |
| Tolerance | 0.05 mL |
| Volume (Metric) | 50 mL |
| Length (Metric) | 540 ±10 mm |