missing translation for 'onlineSavingsMsg'
Learn More

CK 666, Tocris Bioscience™

Product Code. 18085922
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Arp2/3 inhibitor; inhibits actin polymerization

Inhibitor of the Arp2/3 complex; inhibits actin polymerization (IC50 = 4 μM).

Chemical Identifiers

CAS 442633-00-3
Molecular Formula C18H17FN2O
Molecular Weight (g/mol) 296.345
InChI Key UXRKUKRXVWJFER-UHFFFAOYSA-N
Synonym 2-fluoro-n-2-2-methyl-1h-indol-3-yl ethyl benzamide, 2-fluoro-n-2-2-methyl-1h-indol-3-yl-ethyl-benzamide, 3ukr, ck hplc , powder, 2-fluoro-n-2-2-methyl-1h-indol-3-yl ethyl-benzamide, 2-fluorophenyl-n-2-2-methylindol-3-yl ethyl carboxamide, ckh
PubChem CID 589075
ChEBI CHEBI:78843
IUPAC Name 2-fluoro-N-[2-(2-methyl-1H-indol-3-yl)ethyl]benzamide
SMILES CC1=C(C2=CC=CC=C2N1)CCNC(=O)C3=CC=CC=C3F

Specifications

Quantity 50 mg
Formula Weight 296.34
Percent Purity >99%
Chemical Name or Material CK 666
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.