missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Citric Acid, Anhydrous p.a., Thermo Scientific Chemicals
48.74€ - 195.98€
Chemical Identifiers
CAS | 77-92-9 |
---|---|
Molecular Formula | C6H8O7 |
Molecular Weight (g/mol) | 192.12 |
MDL Number | MFCD00011669 |
InChI Key | KRKNYBCHXYNGOX-UHFFFAOYSA-N |
Synonym | citric acid, citric acid, anhydrous, citro, anhydrous citric acid, citrate, aciletten, citretten, chemfill, hydrocerol a, 1,2,3-propanetricarboxylic acid, 2-hydroxy |
PubChem CID | 311 |
ChEBI | CHEBI:30769 |
IUPAC Name | 2-hydroxypropane-1,2,3-tricarboxylic acid |
SMILES | OC(=O)CC(O)(CC(O)=O)C(O)=O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10531471
|
Thermo Scientific
220341000 |
100 g | Plastic bottle |
48.74€
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10551851
|
Thermo Scientific
220345000 |
500 g | Plastic bottle |
60.30€
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11965061
|
Thermo Scientific Acros
220340025 |
2.5 kg | Plastic Bottle |
195.98€
2.50kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Biological buffer for analysisChemical Identifiers
77-92-9 | |
192.12 | |
KRKNYBCHXYNGOX-UHFFFAOYSA-N | |
311 | |
2-hydroxypropane-1,2,3-tricarboxylic acid |
C6H8O7 | |
MFCD00011669 | |
citric acid, citric acid, anhydrous, citro, anhydrous citric acid, citrate, aciletten, citretten, chemfill, hydrocerol a, 1,2,3-propanetricarboxylic acid, 2-hydroxy | |
CHEBI:30769 | |
OC(=O)CC(O)(CC(O)=O)C(O)=O |
Specifications
77-92-9 | |
White | |
C6H8O7 | |
MFCD00011669 | |
Solubility in water: 750g/L (20°C). Other solubilities: soluble in alcohol and ether, badly soluble in diethyl ether, insoluble in benzene and chloroform | |
OC(=O)CC(O)(CC(O)=O)C(O)=O | |
192.12 | |
CHEBI:30769 | |
99.5% | |
100°C | |
0.5% max. | |
Citric acid |
153.0°C | |
99.5% | |
HO2CCH2C(OH)(CO2H)CH2CO2H | |
citric acid, citric acid, anhydrous, citro, anhydrous citric acid, citrate, aciletten, citretten, chemfill, hydrocerol a, 1,2,3-propanetricarboxylic acid, 2-hydroxy | |
KRKNYBCHXYNGOX-UHFFFAOYSA-N | |
2-hydroxypropane-1,2,3-tricarboxylic acid | |
311 | |
192.13 | |
Analytical | |
Authentic | |
Crystals or Crystalline Powder |
Safety and Handling
Warning
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- Citric acid anhydrous
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
Hazard Statement
- H319-Causes serious eye irritation.
Precautionary Statement
- P264-Wash face, hands and anyexposed skin thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P337+P313-If eye irritation persists: Get medical advice/attention.
RUO – Research Use Only