missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Citrate-buffered saline (20X)

32.76€ - 53.27€

Specifications

Concentration 20X
Molecular Weight (g/mol) 294.10
CAS 6132-04-3
Synonym SSC buffer, 20X; Saline-sodium citrate buffer, 20X
Content And Storage Ambient Temperatures
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15415849
View Documents
Thermo Scientific™
J63666.K2
1 L
32.76€
1 L
Please to purchase this item. Need a web account? Register with us today!
15425849
View Documents
Thermo Scientific™
2 L
53.27€
2 L
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

20X
6132-04-3
Ambient Temperatures
294.10
SSC buffer, 20X; Saline-sodium citrate buffer, 20X
Liquid
Videos
SDS
Documents

Documents

Product Certifications