missing translation for 'onlineSavingsMsg'
Learn More

cis-Epoxysuccinic acid, 97%

Product Code. p-3212533 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 10630595

Brand: Thermo Scientific Acros 372640050

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

Currently Unavailable: We have no information when this product will be available.
View alternative products
This item is not returnable. View return policy

CAS: 16533-72-5 | C4H4O5 | 132.07 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16533-72-5
Molecular Formula C4H4O5
Molecular Weight (g/mol) 132.07
MDL Number MFCD00191645
InChI Key DCEMCPAKSGRHCN-XIXRPRMCSA-N
Synonym cis-epoxysuccinic acid, 2r,3s-oxirane-2,3-dicarboxylic acid, cis-oxirane-2,3-dicarboxylic acid, 2,3-oxiranedicarboxylic acid, 2r,3s-rel, 2,3-oxiranedicarboxylic acid, 2r,3s-rel-9ci
PubChem CID 2734802
IUPAC Name (2S,3R)-oxirane-2,3-dicarboxylic acid
SMILES C1(C(O1)C(=O)O)C(=O)O

Specifications

Melting Point 133°C to 135°C
Assay Percent Range 97%
Packaging Glass Bottle
Quantity 5 g
Solubility Information Solubility in water: .. Other solubilities: soluble in organic solvents
Formula Weight 132.07
Percent Purity 97%
Chemical Name or Material cis-Epoxysuccinic acid
compliance-icons
Product Identifier
  • cis-Epoxysuccinic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.