missing translation for 'onlineSavingsMsg'
Learn More
Learn More
cis-4-Hydroxy-D-proline, 98+%, Thermo Scientific Chemicals
34.00€ - 98.50€
Identificadores de productos químicos
CAS | 2584-71-6 |
---|---|
Molecular Formula | C5H9NO3 |
Molecular Weight (g/mol) | 131.13 |
MDL Number | MFCD00005252 |
InChI Key | PMMYEEVYMWASQN-QWWZWVQMSA-N |
Synonym | cis-4-hydroxy-d-proline, h-d-cis-hyp-oh, 2r,4r-4-hydroxypyrrolidine-2-carboxylic acid, d-cis-hydroxyproline, d-allo-hydroxyproline, d-proline, 4-hydroxy-, 4r, 4r-4-hydroxy-d-proline, d-cis-4-hydroxyproline, cis-4-hydroxy-d-prolin, 2r,4r-4-hydroxy-pyrrolidine-2-carboxylic acid |
PubChem CID | 440014 |
ChEBI | CHEBI:16231 |
IUPAC Name | (2R,4R)-4-hydroxypyrrolidine-2-carboxylic acid |
SMILES | O[C@H]1CN[C@H](C1)C(O)=O |
Código de producto | Marca | Quantity | Precio | Cantidad y disponibilidad | |||||
---|---|---|---|---|---|---|---|---|---|
Código de producto | Marca | Quantity | Precio | Cantidad y disponibilidad | |||||
15473737
|
Thermo Scientific Alfa Aesar
H27633.MD |
250 mg |
34.00€
250mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11416590
|
Thermo Scientific Alfa Aesar
H27633.03 |
1 g |
98.50€
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Descripción
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Identificadores de productos químicos
2584-71-6 | |
131.13 | |
PMMYEEVYMWASQN-QWWZWVQMSA-N | |
440014 | |
(2R,4R)-4-hydroxypyrrolidine-2-carboxylic acid |
C5H9NO3 | |
MFCD00005252 | |
cis-4-hydroxy-d-proline, h-d-cis-hyp-oh, 2r,4r-4-hydroxypyrrolidine-2-carboxylic acid, d-cis-hydroxyproline, d-allo-hydroxyproline, d-proline, 4-hydroxy-, 4r, 4r-4-hydroxy-d-proline, d-cis-4-hydroxyproline, cis-4-hydroxy-d-prolin, 2r,4r-4-hydroxy-pyrrolidine-2-carboxylic acid | |
CHEBI:16231 | |
O[C@H]1CN[C@H](C1)C(O)=O |
Especificaciones
243°C (decomposition) | |
C5H9NO3 | |
cis-4-hydroxy-d-proline, h-d-cis-hyp-oh, 2r,4r-4-hydroxypyrrolidine-2-carboxylic acid, d-cis-hydroxyproline, d-allo-hydroxyproline, d-proline, 4-hydroxy-, 4r, 4r-4-hydroxy-d-proline, d-cis-4-hydroxyproline, cis-4-hydroxy-d-prolin, 2r,4r-4-hydroxy-pyrrolidine-2-carboxylic acid | |
O[C@H]1CN[C@H](C1)C(O)=O | |
131.13 | |
CHEBI:16231 | |
≥98% |
2584-71-6 | |
MFCD00005252 | |
PMMYEEVYMWASQN-QWWZWVQMSA-N | |
(2R,4R)-4-hydroxypyrrolidine-2-carboxylic acid | |
440014 | |
131.13 | |
cis-4-Hydroxy-D-proline |
Seguridad y manipulación
missing translation for 'einecsNumber' : 219-963-5
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only