missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ CINPA 1
Potent CAR antagonist
166.30€ - 658.73€
Specifications
Chemical Name or Material | Ethyl [5-[(diethylamino)acetyl]-10,11-dihydro-5H-dibenz[b,f]azepin-3-yl]carbamate |
---|---|
CAS | 102636-74-8 |
Target | Constitutive Androstane Receptor Antagonists |
Molecular Formula | C23H29N3O3 |
Formula Weight | 395.49 |
Description
Potent CAR antagonist (IC50 = 70 nM). Exhibits >90-fold selectivity for CAR over pregnane X receptor (PXR). Also selective over a panel of other nuclear receptors including FXR, LXR, PPARγ and RXR.Specifications
Ethyl [5-[(diethylamino)acetyl]-10,11-dihydro-5H-dibenz[b,f]azepin-3-yl]carbamate | |
Constitutive Androstane Receptor Antagonists | |
395.49 |
102636-74-8 | |
C23H29N3O3 | |
>98% |
Safety and Handling
Recommended Storage : Store at -20°C