missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ CIM 0216

Selective TRPM3 agonist

282.00€ - 1125.00€

Specifications

Chemical Name or Material 3,4-Dihydro-N-(5-methyl-3-isoxazolyl)-α-phenyl-1(2H)-quinolineacetamide
CAS 1031496-06-6
Target TRPM Agonists
Molecular Formula C21H21N3O2
Formula Weight 347.41
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18771598
View Documents
Tocris Bioscience™
5521/50
50 mg
1125.00€
50mg
Please to purchase this item. Need a web account? Register with us today!
18743939
View Documents
Tocris Bioscience™
5521/10
10 mg
282.00€
10mg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Selective TRPM3 agonist. Exhibits selectivity for TRPM3 over TRPM1, TRPM2 and TRPM4-8. Stimulates insulin release from pancreatic islet cells in vitro. Evokes nocifensive behavior in mice.
Specifications

Specifications

3,4-Dihydro-N-(5-methyl-3-isoxazolyl)-α-phenyl-1(2H)-quinolineacetamide
TRPM Agonists
347.41
1031496-06-6
C21H21N3O2
>98%
Videos
Safety and Handling

Safety and Handling

Recommended Storage : Store at 4°C

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.