missing translation for 'onlineSavingsMsg'
Learn More

CIL-102, Thermo Scientific™

Product Code. 15416199
Click to view available options
Quantity:
10 mg
Packaging:
Plastic bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 479077-76-4
Packaging Plastic bottle
Molecular Formula C19H14N2O2
MDL Number MFCD08277043
Quantity 10 mg
Synonym 1-4-furo 2,3-b quinolin-4-ylamino phenyl ethanone, cil hplc , solid, ethanone, 1-4-furo 2,3-b quinolin-4-ylamino phenyl
InChI Key VJDPGTFIMDGXDQ-UHFFFAOYSA-N
SMILES CC(=O)C1=CC=C(C=C1)NC2=C3C=COC3=NC4=CC=CC=C42
IUPAC Name 1-[4-(furo[2,3-b]quinolin-4-ylamino)phenyl]ethanone
Molecular Weight (g/mol) 302.333
PubChem CID 404353
Formula Weight 302.33
Physical Form Powder
Chemical Name or Material CIL-102
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'