Learn More
Proteintech ChromoTek V5-Trap™ Magnetic Agarose
The ChromoTek V5-Trap™ Magnetic Agarose is an affinity resin for IP of V5-tagged proteins.
Brand:  Proteintech v5tma-200
 View more versions of this product
Product Code. 17383543
  • 3237.00€ / 200 reactions
In Stock
to see stock.
Explore more special offers
Description
  • Consists of a anti-V5 Nanobody/VHH coupled to magnetic agarose beads
Specifications
Specifications
Shipped at ambient temperature. Upon receipt store at 4°C. Stable for one year.
V5-tag sequence GKPIPNPLLGLDST at the N-, C-terminal or internal tagging.
Alpaca
IP, CoIP, ChIP, RIP
Magnetic Agarose
200 Reactions