Learn More
Proteintech ChromoTek Spot-Trap™ Magnetic Agarose
Spot-Trap™ Magnetic Agarose is an affinity reagent for immunoprecipitation (IP) of Spot-tagged proteins.
Brand:  Proteintech etma-400
 View more versions of this product
Product Code. 17313323
  • 5899.00€ / 400 reactions
Estimated Shipment: 29-01-2025
to see stock.
Explore more special offers
Description
  • Comprises an anti-Spot-Tag™ VHH/ Nanobody coupled to magnetic agarose beads
Specifications
Specifications
Upon receipt store at 4°C. Do not freeze.
Spot-Tag Sequence Motif PDRVRAVSHWSS
Alpaca
IP, CoIP, ChIP, RIP
Magnetic Agarose
400 Reactions