missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cholesteryl oleyl carbonate, +99%, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
25 g
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Color | White to Yellow |
Infrared Spectrum | Authentic |
Assay Percent Range | 99+% |
Packaging | Glass bottle |
Quantity | 5 g |
Specific Rotation | -22° to -24° (27°C, 589nm) (c=2, CHCl3) |
Formula Weight | 681.14 |
Percent Purity | ≥99.0% |
Physical Form | Waxy Crystalline Mass |
Chemical Name or Material | Cholesteryl oleyl carbonate, 99+% |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'