missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chlorodi(1-naphthyl)phosphine, 95%, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Solubility
Reacts with water.
Notes
Air & Moisture Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, water, moisture, bases.
Reacts with water.
Notes
Air & Moisture Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, water, moisture, bases.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Melting Point | 154°C to 157°C |
Boiling Point | 227°C to 229°C (0.6 mmHg) |
Quantity | 1 g |
UN Number | UN3261 |
Sensitivity | Air and moisture sensitive |
Solubility Information | Reacts with water. |
IUPAC Name | chloro(dinaphthalen-1-yl)phosphane |
Formula Weight | 320.76 |
Percent Purity | 95% |
Chemical Name or Material | Chlorodi(1-naphthyl)phosphine |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'