missing translation for 'onlineSavingsMsg'
Learn More
Learn More
WTW™ Chloride Test Kits
Get accurate water test results for waste water, drinking water, ground water, and mineral water/after with chromate reagent test kits.
121.00€ - 149.00€
Product Code | Brand | Type | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Type | Price | Quantity & Availability | |||||
11785783
|
WTW™
250353 |
Chromate Reagent Test Kit |
121.00€
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10726452
|
WTW™
250491 |
CombiCheck Ammonium (NH4) Reagent Test Kit |
149.00€
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
- Variety of instrumentation for measuring chloride
- Chromate reagent test kit
- For waste water, drinking water, ground water, mineral water/after dilution
- PhotoLab™ S6, PhotoLab™ S12, PhotoLab™ Spectral, Photoflex photometers
Specifications
5 mL |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- Chloride Cell Test Method: photometric 5 - 125 mg/l Cl
Signal Word
- Warning
Hazard Category
- Substances and mixtures corrosive to metals Category 1
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
Hazard Statement
- H290-May be corrosive to metals.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
Precautionary Statement
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
- MIXTURE LIST-Contains : iron(III) nitrate, nitric acid