missing translation for 'onlineSavingsMsg'
Learn More

Kautex™ Wide-Mouth Square Bottles without Closures

Manufactured with HDPE for strength and durability. Kautex™ Wide-Mouth Square Bottles without Closures are ideal for liquids, pastes and granulates.

Brand:  Kautex™ 2000079213

Product Code. 15901441

  • 161.00€ / Pack of 27
In Stock
to see stock.
Edge
Explore more special offers
Add to basket

Description

Description

  • Easy to fill and clean
  • Square cross-section
  • Recessed labeling field
  • Three types of closures available separately
  • UN packaging approved for solids (valid with associated closures)
Application: 
  • packaging
  • sending samples
  • transport
  • storage
TRUSTED_SUSTAINABILITY
Specifications

Specifications

2500 mL
122 x 122 x 260 mm
HDPE
White
68.5 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
View accessory products
Promotions

Promotions