missing translation for 'onlineSavingsMsg'
Learn More

WTW™ Chemical Oxygen Demand (COD) Test Kits

Get accurate Chemical Oxygen Demand (COD) test results with the Chlorine dioxide reagent test kit.

Brand:  WTW™ 251990


 View more versions of this product

Regulatory: This product contains a substance listed in Annex XIV to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery. Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.

Product Code. 11747105

  • 98.40€ / 400mL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Variety of instrumentation for measuring Chemical Oxygen Demand (COD)
  • Safe determinations of COD in water
  • Chlorine dioxide (ClO<sub>2</sub>) reagent test kit
  • For waste water, production control, ground water, surface water
  • For PhotoLab&reg; S6, PhotoLab&reg; S12, PhotoLab&reg; Spectral, spectroFlex photometers
TRUSTED_SUSTAINABILITY
Specifications

Specifications

EN ISO 15705
PhotoLab™ 6000 photometer, Photoflex™ meter
4 to 5 at 25°C
Wastewater, production control, ground water, surface water
100
Fluoride (F) reagent test kit
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • COD Reagent Vario LR

Signal Word
  • Danger

Hazard Category
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 1 A
  • Serious eye damage/eye irritation Category 1
  • Acute toxicity Category 3
  • Specific target organ toxicity after repeated exposure Category 2
  • ACUTE AQUATIC Acute 1
  • LONG-TERM AQUATIC HAZARD Chronic 1

Hazard Statement
  • H290-May be corrosive to metals.
  • H314-Causes severe skin burns and eye damage.
  • H311-Toxic in contact with skin.
  • H373-May cause damage to organs.
  • H410-Very toxic to aquatic life with long lasting effects.
  • H302-Harmful if swallowed.
  • H400-Very toxic to aquatic life.
  • H318-Causes serious eye damage.

Precautionary Statement
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P308+P311-IF exposed or concerned: Call a POISON CENTER/doctor/.

Supplemental information
  • MIXTURE LIST-Contains : sulphuric acid, mercury sulphate, disilver(1+) sulphate, potassium dichromate

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.