missing translation for 'onlineSavingsMsg'
Learn More

CHEMetrics™ Cyanide Neutralizer Solution

Product Code. 11771264
Click to view available options
Quantity:
20 mL
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 11771264

Brand: CHEMetrics™ A3800

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products

This item is not returnable. View return policy

An accessory solution used in conjunction with reagent ampoules in the determination of free cyanide in water

  • Approximately 200 tests
  • 20mL bottles
  • Shelf life: 2 years
  • Range between .1 and .1-1ppm

Specifications

pH Range 4
Volume (Metric) 20 mL
For Use With (Application) Determination of cyanide in water
Quantity 20 mL
Range 0 to 0.1 and 0.1 to 1ppm
Storage Requirements 48.88°C
Volume (English) 0.67 oz.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.