missing translation for 'onlineSavingsMsg'
Learn More

CHEMetrics COD Vials Kits

Contains premeasured reagents for accurate, economical determination of COD

Specifications

Analytes Chemical Oxygen Demand (COD)
For Use With (Application) Premeasured reagents for accurate, economical determination of COD, determination of the oxidizable organic matter content of wastewater
For Use With (Equipment) Digestor block as well as a photometer or spectrophotometer
Test Time 2 hr. digestion
Type COD Vials Kit
View More Specs

Includes: Instructions

Produkte 6
Produktcode Marke Certifications/Compliance Range No. of Tests Includes Preis Menge & Verfügbarkeit  
Produktcode Marke Certifications/Compliance Range No. of Tests Includes Preis Menge & Verfügbarkeit  
11863510
missing translation for 'viewDocuments'
CHEMetrics
K7350S
USEPA accepted 0 to 150 ppm 24 25 vials and instruction book N/A
N/A
11701064
missing translation for 'viewDocuments'
CHEMetrics
K7355
USEPA accepted 0 to 150 ppm 149 150 vials and instruction book N/A
N/A
11873510
missing translation for 'viewDocuments'
CHEMetrics
K7360S
USEPA accepted 0 to 1500 ppm 25 25 vials and instruction book N/A
N/A
11731064
missing translation for 'viewDocuments'
CHEMetrics
K7365
USEPA accepted 0 to 1500 ppm 149 150 vials and instruction book N/A
N/A
11883510
missing translation for 'viewDocuments'
CHEMetrics
K7370S
Not USEPA approved 0 to 15,000 ppm (HR+) 24 25 vials and instruction book N/A
N/A
11741064
missing translation for 'viewDocuments'
CHEMetrics
K7375
Not USEPA approved 0 to 15,000 ppm 98 vials and instruction book N/A
N/A
Beschreibung

Beschreibung

  • Contains everything needed (except distilled water) to perform tests
  • Includes vials and instruction book
  • Comes in cardboard box
  • 2-hour digestion time
  • Dichromate reactor digestion

A fresh reagent ampule blank must be prepared for each series of tests; therefore the number of samples that can be tested with each kit will vary.

Spezifikation

Spezifikation

Chemical Oxygen Demand (COD)
Digestor block as well as a photometer or spectrophotometer
COD Vials Kit
Premeasured reagents for accurate, economical determination of COD, determination of the oxidizable organic matter content of wastewater
2 hr. digestion
Videos
Sicherheit und Handhabung

Sicherheit und Handhabung

missing translation for 'shelfLife' : 1 year

Produktidentifikator
  • COD Vials Kit 0-150 ppm (LR) USEPA-accepted

Signalwort
  • Danger

Gefahrenkategorie
  • Acute toxicity Category 3
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • ACUTE AQUATIC Acute 1
  • Carcinogenicity Category 1B
  • Serious eye damage/eye irritation Category 1
  • Substances and mixtures corrosive to metals Category 1
  • Germ cell mutagenicity Category 1B
  • Oxidising liquid Category 3
  • Reproductive toxicity Category 1B
  • Respiratory sensitiser Category 1A
  • Skin corrosion/irritation Category 1 A
  • Specific target organ toxicity after repeated exposure Category 2
  • Specific target organ toxicity Category 3

Gefahrenhinweis
  • H272-May intensify fire; oxidiser.
  • H290-May be corrosive to metals.
  • H302-Harmful if swallowed.
  • H311-Toxic in contact with skin.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H335-May cause respiratory irritation.
  • H340-May cause genetic defects.
  • H350-May cause cancer.
  • H360FD-May damage fertility. May damage the unborn child.
  • H373-May cause damage to organs.
  • H400-Very toxic to aquatic life.
  • H412-Harmful to aquatic life with long lasting effects.
  • EUH208-Contains . May produce an allergic reaction.

Sicherheitshinweis
  • P201-Obtain special instructions before use.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P271-Use only outdoors or in a well-ventilated area.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P403+P233-Store in a well-ventilated place. Keep container tightly closed.
  • P405-Store locked up.
  • P501-Dispose of contents/container to .

Ergänzende Informationen
  • MIXTURE LIST-Contains : Sulfuric acid, Silver sulfate, Mercuricsulfate, Potassium dichromate

Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate