missing translation for 'onlineSavingsMsg'
Learn More

CGP 7930, Tocris Bioscience™

Product Code. 30001262
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Positive allosteric modulator of GABAB receptors

Positive allosteric modulator of GABAB receptors. Increases the potency and efficacy of GABA at both native and recombinant GABAB receptors (EC50 values are 5.37 and 4.60μM respectively) and enhances the inhibitory effect of the agonist L-baclofen in cultured cortical neurons. Reduces operant self-administration of ethanol in alcohol-preferring rats following i.p. administration.

Chemical Identifiers

CAS 57717-80-3
Molecular Formula C19H32O2
Molecular Weight (g/mol) 292.463
InChI Key XLWJPQQFJNGUPA-UHFFFAOYSA-N
Synonym 2,6-di-tert-butyl-4-3-hydroxy-2,2-dimethylpropyl phenol, 2,6-ditert-butyl-4-3-hydroxy-2,2-dimethylpropyl phenol, tocris-1513, lopac-c-0862, cgp hplc , solid, 2,2-di-methyl-3-3,5-di-t-butyl-4-hydroxyphenyl propanol, 3-3,5-di-t-butyl-4-hydroxyphenyl-2,2-dimethylpropanol, 3,5-bis 1,1-dimethylethyl-4-hydroxy-?,?-dimethyl-benzenepropanol, 3-3',5'-di-tert-butyl-4'-hydroxy phenyl-2,2-dimethylpropanol, 3-3 inverted exclamation marka,5 inverted exclamation marka-di-tert-butyl-4 inverted exclamation marka-hydroxy phenyl-2,2-dimethylpropanol
PubChem CID 5024764
IUPAC Name 2,6-ditert-butyl-4-(3-hydroxy-2,2-dimethylpropyl)phenol
SMILES CC(C)(C)C1=CC(=CC(=C1O)C(C)(C)C)CC(C)(C)CO

Specifications

Quantity 10 mg
Formula Weight 292.46
Percent Purity >98%
Chemical Name or Material CGP 7930
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'