missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CGP 52432, Tocris Bioscience™
Click to view available options
Quantity:
10 mg
50 mg
Description
Potent, selective GABAB receptor antagonist (IC50 = 85 nM).
Specifications
Specifications
Quantity | 10 mg |
Formula Weight | 384.24 |
Percent Purity | >99% |
Chemical Name or Material | CGP 52432 |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'