missing translation for 'onlineSavingsMsg'
Learn More

Cevimeline hydrochloride, Tocris Bioscience™

Product Code. 18732774
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Selective M1 agonist

Selective M1 receptor agonist. Induces atropine-sensitive contractions of isolated guinea pig ilea and trachea preparations (EC50 values are 3.5 and 3 μM respectively). Reverses AF64A-induced cognitive impairments in vivo.

Chemical Identifiers

CAS 107220-28-0
Molecular Formula C10H18ClNOS
Molecular Weight (g/mol) 235.77
InChI Key SURWTGAXEIEOGY-UHFFFAOYSA-N
Synonym hydrogen cevimeline chloride
PubChem CID 9837584
IUPAC Name 2-methylspiro[1,3-oxathiolane-5,3'-1-azabicyclo[2.2.2]octane];hydrochloride
SMILES CC1OC2(CN3CCC2CC3)CS1.Cl

Specifications

Quantity 10 mg
Formula Weight 235.77
Percent Purity >98%
Chemical Name or Material Cevimeline hydrochloride
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'