Learn More
BUCHI Ceramic Crucible
For use with B-440 Wet Digesters to aid in the ecological pre-incineration of different types of samples.
Brand: BUCHI 026938
Additional Details : Weight : 0.05000kg
Description
- 90mL capacity
- White in color
- Round in shape
Specifications
White | |
Pre-incineration of samples | |
Crucible |
Ceramic | |
90 mL | |
Round |