Learn More
Labconco™ CentriVap™ Complete Concentrator
Comes with everything needed to start processing
Brand:  Labconco™ 7315060
 View more versions of this product
Product Code. 15223423
Description
Description
Comes with everything needed to start processing
Uses a combination of centrifugal force, vacuum and heat to speed evaporation.
Includes:
  • Vacuum Concentrator
  • -50°C Cold Trap
  • Aluminum rotor holds 40 microcentrifuge tubes and 12-13mm tubes
  • Chemical-resistant diaphragm vacuum pump
  • CentriZap Strobe Light allows user to view samples while rotor is spinning and lid is closed
Molecular biology, proteomics, genomics, cell biology, microbiology, biochemistry, drug discovery and analytical chemistry
Specifications
Specifications
Centrivap
International/UK
Heat Boost Not Included
230 V 50 Hz 6 A