Learn More
Kartell™ Centrifuge Micro Test Tube with Cap
Manufactured of high transparency polypropylene for easy sample checking. Kartell™ Centrifuge Micro Test Tube with Cap feature a large frosted writing surface to improve tracability in sample analysis.
Brand:  Kartell™ 2940
 View more versions of this product
Product Code. 10044722
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products »
Description
Description
  • High transparency PP tube for easy sample checking
  • Flat cap with large frosted writing surface
  • Liquid tight for centrifugation up to RCF 21.000 x g
  • Large opening tab featuring rounded surfaces for smoother open/close routine
  • Cap top with 3mm pierceable area
  • Autoclavable at 121°C for 20 minutes with open cap
Specifications
Specifications
Clear
Cobas Bio and Cobas Mira automatic analyzers
1000/Pk.