missing translation for 'onlineSavingsMsg'
Learn More

OHAUS™ Centrifuge Bucket Accessory

Accessory for centrifuges

Brand:  OHAUS™ 30553104

 View more versions of this product

Product Code. 16692754

  • 1190.00€ / Pack of 2

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Accessory for centrifuges
    • Centrifuge Multi FC5916R 120V
    • Centrifuge Multi FC5916 120V
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Centrifuge Multi FC5916R 120 V and Centrifuge Multi FC5916 120 V
MTP x2 V1
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions