missing translation for 'onlineSavingsMsg'
Learn More
Learn More
OHAUS™ Centrifuge Bucket Accessory
Accessory for centrifuges
Brand: OHAUS™ 30553119
This item is not returnable.
View return policy
Description
- Accessory for centrifuges
- Centrifuge Multi FC5916R 120V
- Centrifuge Multi FC5916 120V
Specifications
Centrifuge Multi FC5916R 120 V and Centrifuge Multi FC5916 120 V | |
MTP x2 V2 |