Learn More
Eppendorf™ Centrifuge 5910 R High Speed Package (EU)
GREENER_CHOICE
Centrifuge 5910 R features advanced temperature management and greater flexibility than standard multipurpose centrifuges.
Brand:  Eppendorf™ 5942000148
Product Code. 16612842
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products »
Explore more special offers
Description
Description
  • High Speed Package
  • With Rotor FA-6x50
  • Refrigerated
  • Aerosol-tight Eppendorf QuickLock™ caps and lids allow fast and ergonomic lid locking
  • Centrifuge lid with soft-touch lid closure
  • Quiet operation to improve your work environment
  • Compact footprint saves valuable bench space
  • Automatic rotor recognition and imbalance detection for maximum operational safety
TRUSTED_SUSTAINABILITY
Specifications
Specifications
22,132 x g
14.5 in.
28.3 in.
1650 w
230 V 50/60 Hz
99 (5 program keys)
Large, Bright LCD
10 sec. to 99 hr. 59 min., with continuous run function, short-spin
72 cm
UL (120 V), CAN/CSA (120 V), CE, RoHS
100 to 14,000 rpm
10/10