Learn More
Eppendorf™ Centrifuge 5425/5425 R
GREENER_CHOICE
Eppendorf’s newest 24-place microcentrifuges, Centrifuge 5425 and 5425 R, feature ergonomic operation and a large selection of rotors with a maximum speed of up to 21,300 x g, making them perfectly suited for all modern molecular biological applications in Eppendorf and PCR tubes from 0.2 to 5 mL.
2375.00€ - 6571.18€
Specifications
Max. RCF 21,300 x g
View More Specs
Products 21
Product Code Brand Model Refrigerated Rotor Included Electrical Requirements Price Quantity & Availability  
Product Code Brand Model Refrigerated Rotor Included Electrical Requirements Price Quantity & Availability  
15831645
GREENER_CHOICE
View accessory products View Documents
Eppendorf™
5405000417
Centrifuge 5425 (EU-IVD), rotary knobs No Yes 230V/50–60Hz (EU) This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
N/A
15841645
GREENER_CHOICE
View accessory products View Documents
Eppendorf™
5405000468
Centrifuge 5425 (EU-IVD), rotary knobs No Yes 230V/50–60Hz (GB) This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
N/A
15851635
GREENER_CHOICE
View accessory products View Documents
Eppendorf™
5405000018
Centrifuge 5425 (EU-IVD), keypad No Yes 230V/50–60Hz (EU) This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
N/A
15861635
GREENER_CHOICE
View accessory products View Documents
Eppendorf™
5405000069
Centrifuge 5425 (EU-IVD), keypad No Yes 230V/50–60Hz (GB) This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
N/A
15871645
GREENER_CHOICE
View accessory products View Documents
Eppendorf™
5405000611
Centrifuge 5425 (EU-IVD), rotary knobs No No 230V/50–60Hz (EU)
2500.00€
Each
15891635
GREENER_CHOICE
View accessory products View Documents
Eppendorf™
5405000212
Centrifuge 5425 (EU-IVD), keypad No No 230V/50–60Hz (EU)
2500.00€
Each
15811645
GREENER_CHOICE
View accessory products View Documents
Eppendorf™
5405000310
Centrifuge 5425, keypad No No 230V/50–60Hz (EU)
2479.88€
Each
15851645
GREENER_CHOICE
View accessory products View Documents Promotion Details
Eppendorf™
5405000514
Centrifuge 5425, rotary knobs No FA-24x2 230V/50–60Hz (EU) This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
N/A
15861645
GREENER_CHOICE
View accessory products View Documents
Eppendorf™
5405000565
Centrifuge 5425, rotary knobs No Yes 230V/50–60Hz (GB) This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
N/A
15871635
GREENER_CHOICE
View accessory products View Documents
Eppendorf™
5405000115
Centrifuge 5425, keypad No Yes 230V/50–60Hz (EU) This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
N/A
15881635
GREENER_CHOICE
View accessory products View Documents
Eppendorf™
5405000166
Centrifuge 5425, keypad No Yes 230V/50–60Hz (GB) This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
N/A
15891645
GREENER_CHOICE
View accessory products View Documents
Eppendorf™
5405000719
Centrifuge 5425, rotary knobs No No 230V/50–60Hz (EU)
2375.00€
Each
16855100
GREENER_CHOICE
View accessory products View Documents Promotion Details
Eppendorf™
5406000119
Centrifuge 5425 R, keypad Yes Yes 230V/50–60Hz (EU)
6259.94€
1 set
16895100
GREENER_CHOICE
View accessory products View Documents
Eppendorf™
5406000313
Centrifuge 5425 R, keypad Yes No 230V/50–60Hz (EU)
5585.00€
Each
16835110
GREENER_CHOICE
View accessory products View Documents Promotion Details
Eppendorf™
5406000518
Centrifuge 5425 R, rotary knobs Yes Yes 230V/50–60Hz (EU)
5995.00€
1 set
16875110
GREENER_CHOICE
View accessory products View Documents
Eppendorf™
5406000712
Centrifuge 5425 R, rotary knobs Yes No 230V/50–60Hz (EU)
5585.00€
Each
16835100
GREENER_CHOICE
View accessory products View Documents
Eppendorf™
5406000011
Centrifuge 5425 R (EU-IVD), keypad Yes Yes 230V/50–60Hz (EU)
6571.18€
1 set
16875100
GREENER_CHOICE
View accessory products View Documents
Eppendorf™
5406000216
Centrifuge 5425 R (EU-IVD), keypad Yes No 230V/50–60Hz (EU)
5865.00€
Each
16815110
GREENER_CHOICE
View accessory products View Documents
Eppendorf™
5406000410
Centrifuge 5425 R (EU-IVD), rotary knobs Yes Yes 230V/50–60Hz (EU)
6305.00€
1 set
16855110
GREENER_CHOICE
View accessory products View Documents
Eppendorf™
5406000615
Centrifuge 5425 R (EU-IVD), rotary knobs Yes No 230V/50–60Hz (EU)
5865.00€
Each
17474152
GREENER_CHOICE
View Documents Promotion Details
Eppendorf™
5405000158
Centrifuge 5425 (EU-IVD) No Yes 230V/50–60Hz (EU)
2825.00€
1 set
Description
Description
For processing temperature-sensitive samples, the refrigerated Centrifuge 5425 R provides precise temperature control with its advanced cooling system.
  • Aerosol-tight Eppendorf QuickLock™ lids for simple and safe handling
  • Outstanding software features (e.g. adjustable acceleration and braking ramps, 3 program buttons (only Centrifuge 5425), short function without continuous pressing of the key)
  • Extended rotor program (e.g. 5.0 mL rotor, swing-bucket rotor for PCR strips)
  • OptiBowl™ design for quiet operation, even without a rotor lid
  • Soft-touch one-finger lid closure for ergonomic operation
  • Max. rotor capacity: 10 × 5.0 mL vessels
  • Max. speed: 21,300 × g (15,060 rpm)
Specifications
Specifications
21,300 x g
Documents
Product Certifications
The Centrifuge 5425 is designed for separating liquid substance mixtures with different densities, in particular, for processing and analyzing samples from the human body in in-vitro diagnostic applications to ensure that the in-vitro diagnostic device can be used according to its intended purpose. This centrifuge including components is an in-vitro diagnostic device according to Directive 98/79/EC of the European Parliament and the Council dated October 27, 1998. Eppendorf centrifuges are exclusively intended for indoor use by trained specialists.
Product Content Correction
Product Title
Eppendorf™ Centrifuge 5425/5425 R