Learn More
Greiner Bio-One CELLSTAR Suspension Cell Culture Flasks
Designed to culture cell suspended in a growth medium. Greiner Bio-One™ CELLSTAR Suspension Cell Culture Flasks are manufactured from polystyrene for optimum visibility and strength.
Brand:  Greiner Bio-One 690190
 View more versions of this product
Product Code. 10722541
  • 457.00€ / Pack of 200
Estimated Shipment: 27-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Hydrophobic surface, ideal for suspension cultures and hybridoma
  • Free of detectable DNase, RNase, human DNA
  • Non-Pyrogenic, non-cytotoxic
  • Graduation on the side
  • Canted neck
  • Easily distinguished by their white screw caps
  • White standard screw cap with ventilation position
  • TC surface treatment: -/suspension
  • Sterile and user friendly packaging
TRUSTED_SUSTAINABILITY
Specifications
Specifications
White
Polystyrene
200
Screw
50 mL
200 Pack
Yes