Learn More
Greiner Bio-One CELLSTAR μClear™ 96-well, Cell Culture-Treated, Flat-Bottom, Half-Area Microplate
Color:Black, Clear Bottom; Sterility:Sterile; Lid:With Lid
Brand: Greiner Bio-One 675090
This item is not returnable.
View return policy
Description
Alphanumeric codingSpecifications
Half-Area | |
8 x 12 | |
Black, Clear Bottom | |
96 | |
Sterile | |
199 μL | |
Yes | |
32 | |
Yes | |
127.76 x 85.48 x 14.40 mm |
With Lid | |
Not Autoclavable | |
Polystyrene | |
15 to 175 μL | |
Cell Culture | |
Round, Flat Bottom (F-well) | |
4800 x g (swinging bucket rotor) | |
8 | |
Black |