Learn More
Invitrogen™ CellSensor™ TrkB-NFAT-bla CHO-K1 Cell Line
Product Code. 10236844
Click to view available options
Quantity:
1 vial
Unit Size:
Each
Product Code. 10236844
Brand: Invitrogen™ K1491
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
CellSensor™ TrkB-NFAT-bla CHO-K1 Cell Line
Neurotrophins (NGF, BDNF, NT-3, and NT-4) and their transmembrane receptors (TrkA, TrkB, TrkC, and P75NTR) play important roles in the regulation of neuronal and non-neuronal cell proliferation, differentiation, survival, and death. Neurotrophin signaling also mediates higher-order neuronal activities, such as learning, memory, and behavior. Alterations in neurotrophin levels and their receptors have been implicated in neurodegenerative disorders, such as Alzheimer's disease and Huntington's disease, as well as psychiatric disorders. Neurotrophins propagate their signal through activating multiple signaling pathways (e.g., MAPK, PI3K, PKC).
BDNF (brain-derived neurotrophic factor) and its receptor TrkB (also known as NTRK2) play a fundamental role in regulating neural development, survival, and synaptic activity and plasticity. TrkB is also a potential anticancer target, since altered signaling through TrkB promotes tumor formation, survival, and metastasis of various cancers (including neuroblastomas, multiple myelomas, and pancreatic ductal adenocarcinomas). Moreover, growing evidence indicates that BDNF and its receptor influences food intake and body weight control.
CellSensor™ TrkB-NFAT-bla CHO-K1 cells contain a beta-lactamase reporter gene under control of the NFAT Response Element that has been stably integrated into CHO-K1 cells along with TrkB. CellSensor™ TrkB-NFAT-bla CHO-K1 cells express beta-lactamase upon stimulation with brain-derived neurotrophic factor (BDNF). This cell line is a clonal population isolated by flow cytometry and has been tested for robust performance by assessing a variety of assay parameters.
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage CellSensor™ TrkB-NFAT-bla CHO-K1 cells are shipped on dry ice. Store in liquid nitrogen immediately upon receipt, or thaw for immediate use.
Assay Entry Cell-based beta lactamase reporter gene
Target Entry TRKB, PKC and calcium pathway
Quantity 1 vial
Product Type TrkB-NFAT-bla CHO-K1 Cell Line
Product Line CellSensor