Learn More
Thermo Scientific™ CellSensor™ T-REx™ NICD CSL-bla HeLa Cell Line
This cell line is a clonal population isolated by flow cytometry. Addition of doxycycline to these cells allows for regulated NICD transcription factor expression and subsequent beta-lactamase expression.
Brand:  Thermo Scientific™ K1487
Product Code. 10607363
  • 11500.00€ / Each
Estimated Shipment: 31-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
The CellSensor™ T-REx™ NICD CSL-bla HeLa cell line was engineered by lentiviral transduction of HeLa cervical cancer cells with a notchresponse element driving beta-lactamase reporter gene expression (CSL-bla) along with tetracycline repressor and tetracycline (or the tetracycline analog, doxycycline)-inducible NICD (notch intracellular domain) constructs. This cell line is a clonal population isolated by flow cytometry. Addition of doxycycline to these cells allows for regulated NICD transcription factor expression and subsequent beta-lactamase expression. This cell line has been tested for robust assay performance by assessing a variety of parameters, including cell plating number, DMSO tolerance, stimulation time, and substrate loading time. Assay validation was performed using serial dilutions of doxycycline. Additional testing data using RNAi and alternate stimuli are also available.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Cell-based beta lactamase reporter gene
CellSensor™ T-REx™ NICD CSL-bla HeLa cells are shipped on dry ice. Store in liquid nitrogen immediately upon receipt, or thaw for immediate use.
Signaling Pathway
1 Vial
Dividing Cells
CellSensor™
Dividing Cells
Fluorescence
CellSensor™, T-REx™
NICD, Notch pathway
HeLa
Signaling Pathway
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
For Research Use Only. Not for use in diagnostic procedures.