Learn More
Invitrogen™ CellSensor™ Myc-bla HCT116 Cell Line
CellSensor™ Myc-bla HCT116 Cell Line
Brand: Invitrogen™ K1467
This item is not returnable.
View return policy
Description
The CellSensor™Myc-bla HCT116 cell line contains a beta-lactamase reporter gene under the control of Myc binding sequences. The construct was transduced into HCT116 cells using a lentiviral system. HCT116 is a colon cancer cell line which expresses a mutated form of beta-catenin. This form of beta-catenin leads to the accumulation of beta-catenin and constitutive activation of downstream genes such as Myc. This cell line is a clonal population isolated by flow cytometry. It has been validated for cell plating density and DMSO tolerance. The signaling pathway has been validated using RNAi against c-Myc and ICG-001, an inhibitor of the wnt-beta-catenin pathway. The expression of the mutant beta-catenin in HCT116 cells results in constitutive activation of beta-lactamase in this CellSensor™line, which can be knocked down by ICG-001 (Figure 1) or Myc RNAi (Figure 2).Specifications
Cell-based beta lactamase reporter gene | |
CellSensor™ Myc-bla HCT116 cells are shipped on dry ice. Store in liquid nitrogen immediately upon receipt, or thaw for immediate use. |
|
CellSensor™ | |
Myc pathway, MYC | |
HCT-116 | |
Signaling Pathway |
Dividing Cells | |
Fluorescence | |
1 vial | |
Cell Line | |
CellSensor™ |
For Research Use Only. Not for use in diagnostic procedures.