Learn More
Invitrogen™ CellSensor™ Myc-bla HCT116 Cell Line
CellSensor™ Myc-bla HCT116 Cell Line
Brand:  Invitrogen™ K1467
Product Code. 10247224
  • 11390.00€ / Each
Estimated Shipment: 29-11-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
The CellSensor™Myc-bla HCT116 cell line contains a beta-lactamase reporter gene under the control of Myc binding sequences. The construct was transduced into HCT116 cells using a lentiviral system. HCT116 is a colon cancer cell line which expresses a mutated form of beta-catenin. This form of beta-catenin leads to the accumulation of beta-catenin and constitutive activation of downstream genes such as Myc. This cell line is a clonal population isolated by flow cytometry. It has been validated for cell plating density and DMSO tolerance. The signaling pathway has been validated using RNAi against c-Myc and ICG-001, an inhibitor of the wnt-beta-catenin pathway. The expression of the mutant beta-catenin in HCT116 cells results in constitutive activation of beta-lactamase in this CellSensor™line, which can be knocked down by ICG-001 (Figure 1) or Myc RNAi (Figure 2).
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Cell-based beta lactamase reporter gene
CellSensor™ Myc-bla HCT116 cells are shipped on dry ice.
Store in liquid nitrogen immediately upon receipt, or thaw for immediate use.
CellSensor™
Myc pathway, MYC
HCT-116
Signaling Pathway
Dividing Cells
Fluorescence
1 vial
Cell Line
CellSensor™
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
For Research Use Only. Not for use in diagnostic procedures.