Learn More
Thermo Scientific™ CellSensor™ GAS-bla ME-180 Cell Line
The CellSensor™ GAS-bla ME-180 cell line contains a beta-lactamase reporter gene under control of the Gamma Activated Sequence (GAS) response element stably integrated into ME-180 cells.
Brand:  Thermo Scientific™ K1651B
Product Code. 10308283
  • 11350.00€ / Each
Estimated Shipment: 24-01-2025
to see stock.
Explore more special offers
Description
To construct this cell line, the GAS-bla construct was transduced into ME-180 cells by lentivirus. Flow cytometry was used to isolate cells responsive to Interferon Gamma (IFN-γ). This cell line is validated for DMSO tolerance, cell number, stimulation time, substrate loading time, as well as Z'and EC50 concentrations for IFN-γ. The CellSensor™ GAS-bla ME-180 cell line is responsive to IFN-γ and can be used to probe the JAK/STAT signaling pathway. This cell line can be adapted for high-throughput screening of agonist or antagonist compound libraries. Candidate drugs can be tested for dose response against this cell line. Academic and non-profit customers, please inquire for special pricing.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Dividing Cells
1 vial
Signaling Pathway
CellSensor™ Cell Lines are shipped on dry ice. Store in liquid nitrogen immediately upon receipt, or thaw for immediate use.
CellSensor™