missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CellBlox™ Blocking Buffer

Product Code. 17276356
Click to view available options
Quantity:
100 Tests
25 Tests
500 Tests
Unit Size:
Pack of 100
Pack of 25
Pack of 500
This item is not returnable. View return policy

Product Code. 17276356

Brand: Invitrogen™ B001T03F01

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products

This item is not returnable. View return policy

Formulated to block nonspecific binding of Invitrogen™ NovaFluor™ labels with cells, which can result in higher background labeling. Also recommended for cyanine-based dyes or cyanine-based tandem dyes to block non-specific interactions with monocytes and macrophages to minimize background labeling.

  • Formulated to block nonspecific binding of Invitrogen™ NovaFluor™ labels with cells that can result in higher background labeling
  • CellBlox™ Blocking Buffer is a non-antibody, non-protein–based blocking solution, and should be used every time a NovaFluor dye is used for labeling any cell type to minimize background labeling
  • Recommended for use with cyanine-based dyes or cyanine-based tandem dyes to block non-specific interactions with monocytes and macrophages to minimize background labeling
  • Use of CellBlox Blocking Buffer requires minimal change to most flow cytometry staining protocols
  • Add 5 μL of buffer directly to a cell suspension containing 103 to 108 cells prior to the addition of an antibody, with 100 μL as a final staining volume
  • Buffer may be added to an antibody cocktail mixture prior to labeling cells, by adding 5 μL of buffer for every stained sample to be labeled with the antibody cocktail mixture, with 100 μL as a final staining volume
  • Always use CellBlox Blocking Buffer with NovaFluor dyes when labeling cells for best background reduction
  • Compatible with all fluorophores and with Invitrogen™ LIVE/DEAD™ Fixable Dead Cell Stains
  • Can be used with any fluorophore-antibody conjugate as a high-performance monocyte and macrophage blocking solution
  • CellBlox Blocking Buffer is compatible with other blocking reagents such as Fc Block, blocking proteins, Brilliant Stain Buffer, and Super Bright Complete Staining Buffer
  • Buffer is not required when labeling antibody-capture beads
TRUSTED_SUSTAINABILITY

Specifications

Description Blocking Buffer
For Use With (Application) Flow Cytometry
Quantity 100 Tests
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.