Learn More
Invitrogen™ eBioscience™ Cell Stimulation Cocktail (plus protein transport inhibitors) (500X)
Can be used for induction and subsequent intracellular detection of cytokines and other secreted proteins in both in vitro-cultured and ex vivo cells
125.00€ - 369.00€
Specifications
For Use With (Application) | Flow Cytometry, Functional Assays, Cell Culture |
---|---|
Product Line | eBioscience™ |
Product Type | Cell Stimulation Cocktail |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15516286
|
Invitrogen™
00-4975-93 |
100 μL |
125.00€
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15506286
|
Invitrogen™
00-4975-03 |
4 x 100 μL |
369.00€
Pack of 4 |
Estimated Shipment: 16-12-2024
Log in to see stock. |
|||||
Description
Cocktail of phorbol 12-myristate 13-acetate (PMA), ionomycin, brefeldin A and monensin. Treatment with PMA and ionomycin is sufficient to induce activation of many cell types to produce cytokines. Brefeldin A and Monensin result in the accumulation of secreted proteins in the endoplasmic reticulum and Golgi apparatus
- A cocktail of phorbol 12-myristate 13-acetate (PMA), ionomycin, brefeldin A and monensin
- Treatment with PMA and ionomycin is sufficient to induce activation of many cell types to produce cytokines
- Brefeldin A and Monensin result in the accumulation of secreted proteins in the endoplasmic reticulum and Golgi apparatus
- Thus, it can be used for induction and subsequent intracellular detection of cytokines and other secreted proteins in both in vitro-cultured and ex vivo cells
Specifications
Flow Cytometry, Functional Assays, Cell Culture | |
eBioscience™ | |
Cell Stimulation Cocktail |
For Research Use Only. Not for use in diagnostic procedures.