Learn More
Tocris Bioscience™ CE3F4
Noncompetitive Epac1 inhibitor
Brand: Tocris Bioscience™ 4793/50
This item is not returnable.
View return policy
Description
CE3F4 is a noncompetitive Epac1 inhibitor. Blocks Epac-induced Rap activation and prevents isoprenaline -induced autophagy flux in cardiomyocytes. Has no effect on PKA activity in the presence of cAMP.Specifications
>98% | |
CE3F4 | |
EPAC Inhibitors |
50 mg | |
351.01 |