Learn More
Tocris Bioscience™ CE3F4
Noncompetitive Epac1 inhibitor
Brand:  Tocris Bioscience™ 4793/50
 View more versions of this product
Product Code. 18758748
  • 785.00€ / 50mg
Explore more special offers
Description
Description
CE3F4 is a noncompetitive Epac1 inhibitor. Blocks Epac-induced Rap activation and prevents isoprenaline -induced autophagy flux in cardiomyocytes. Has no effect on PKA activity in the presence of cAMP.
Specifications
Specifications
>98%
CE3F4
EPAC Inhibitors
50 mg
351.01