missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD45.1 Mouse, PE, Clone: A20, enQuireBio™
Mouse Monoclonal Antibody
125.00€ - 250.00€
Specifications
Antigen | CD45.1 |
---|---|
Clone | A20 |
Concentration | 0.5mg/mL |
Applications | Flow Cytometry |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15932018
|
EnquireBio
QAB42-PE-25ug |
25 μg |
125.00€
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15922018
|
EnquireBio
QAB42-PE-100ug |
100 μg |
250.00€
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Specifications
CD45.1 | |
0.5mg/mL | |
Monoclonal | |
Mouse | |
10 mM NaH2PO4, 150 mM NaCl, 0.09% NaN3, 0.1% gelatin, pH7.2 | |
19264 | |
IgG2a κ | |
Protein A/G Purified |
A20 | |
Flow Cytometry | |
PE | |
RUO | |
NP_001255215 | |
Ptprc | |
Primary | |
Store at 2-4°C. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title