missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD34 Antibody (HPCA1/1806R), Alexa Fluor™ 488, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-54355AF488
This item is not returnable.
View return policy
Description
CD34 Monoclonal antibody specifically detects CD34 in Human, Rat samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin), CyTOF
Specifications
CD34 | |
Monoclonal | |
Alexa Fluor 488 | |
CD34 antigenhematopoietic progenitor cell antigen CD34, CD34 molecule | |
Recombinant full-length human CD34 protein (Uniprot: P28906) | |
100 μg | |
Adaptive Immunity, Angiogenesis, B Cell Development and Differentiation Markers, Breast Cancer, Cancer, Cell Biology, Cellular Markers, Endothelial Cell Markers, Hematopoietic Stem Cell Markers, Hypoxia, Immunology, Innate Immunity, Mast Cell Markers, Mesenchymal Stem Cell Markers, Myeloid Cell Markers, Myeloid derived Suppressor Cell, Neuronal Cell Markers, Neuroscience, Stem Cell Markers, Tumor Suppressors | |
947 | |
Store at 4°C in the dark. | |
IgG |
Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin), CyTOF | |
HPCA1/1806R | |
50 mM Sodium Borate | |
Rabbit | |
Protein A or G purified | |
RUO | |
Primary | |
Human, Rat | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
CD34 Antibody (HPCA1/1806R), Alexa Fluor™ 488, Novus Biologicals™ > 100 μg, Alexa Fluor 488
Spot an opportunity for improvement?Share a Content Correction