missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD3 Mouse, PE, Clone: OKT3, enQuireBio™
Mouse Monoclonal Antibody
85.85€ - 162.00€
Specifications
Antigen | CD3 |
---|---|
Clone | OKT3 |
Concentration | 5μL/Test |
Applications | Flow Cytometry |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15922138
|
EnquireBio
QAB5-PE-25Tests |
25 Tests |
85.85€
25 tests |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15912138
|
EnquireBio
QAB5-PE-100Tests |
100 Tests |
162.00€
100 tests |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Specifications
CD3 | |
5μL/Test | |
Monoclonal | |
Mouse | |
10 mM NaH2PO4, 150 mM NaCl, 0.09% NaN3, 0.1% gelatin, pH7.2 | |
916 | |
IgG2a κ | |
Protein A/G Purified |
OKT3 | |
Flow Cytometry | |
PE | |
RUO | |
NP_000724 | |
CD3E | |
Primary | |
Store at 2-4°C. |