missing translation for 'onlineSavingsMsg'
Learn More

CD24 Antibody (SN3) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody has been used in 2 publications

222.00€ - 517.00€

Specifications

Antigen CD24
Clone SN3
Concentration 1.0 mg/mL
Dilution Flow Cytometry 1/50-1/100, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen 1:10-1:500
Applications Immunohistochemistry (Frozen), Immunohistochemistry, Immunohistochemistry (Paraffin), Flow Cytometry
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18027433
View Documents
Novus Biologicals
NB100-64861-0.025MG
0.025 mg
222.00€
25µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18053904
View Documents
Novus Biologicals
NB100-64861
0.1 mg
517.00€
0.10mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

CD24 Monoclonal specifically detects CD24 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.
Specifications

Specifications

CD24
1.0 mg/mL
Immunohistochemistry (Frozen), Immunohistochemistry, Immunohistochemistry (Paraffin), Flow Cytometry
Unconjugated
Mouse
Cancer, Neuronal Cell Markers, Neuronal Stem Cell Markers, Stem Cell Markers
P25063
100133941
Glycoproteins purified from human NALM-1 cell line
Primary
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
SN3
Flow Cytometry 1/50-1/100, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen 1:10-1:500
Monoclonal
Purified
RUO
Human
CD 24, CD24 antigen, CD24 antigen (small cell lung carcinoma cluster 4 antigen), CD24 molecule, CD24Asignal transducer CD24, FLJ22950, FLJ43543, MGC75043, Small cell lung carcinoma cluster 4 antigen
CD24
IgG1
Protein G purified
Recognizes the human CD24 cell surface antigen, a heavily glycosylated molecule of 35-45kD. CD24 is expressed by granulocytes and B lymphocytes, and by some activated T cells and T cell malignancies.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.