missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD11c (Dendritic Cell Marker) Mouse, Clone: ITGAX/2507, enQuireBio™
Mouse Monoclonal Antibody
199.00€ - 478.00€
Specifications
| Antigen | CD11c (Dendritic Cell Marker) |
|---|---|
| Clone | ITGAX/2507 |
| Concentration | 200μg/mL |
| Applications | Immunohistochemistry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
15910298
|
EnquireBio
3687-MSM6-P0 |
20 μg | |||||||
|
15920298
|
EnquireBio
3687-MSM6-P1 |
100 μg |
554.40€ 478.00€ / 100µg Save 76.40€ 13% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Specifica
| CD11c (Dendritic Cell Marker) | |
| 200μg/mL | |
| Monoclonal | |
| Mouse | |
| 10mM PBS with 0.05% BSA and 0.05% azide. Also available without BSA and Azide. | |
| Recombinant fragment of human ITGAX protein (apprx. aa 637-827) | |
| Primary | |
| Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. | |
| 145kDa |
| ITGAX/2507 | |
| Immunohistochemistry | |
| Unconjugated | |
| RUO | |
| 3687 | |
| IgG1 κ | |
| Protein A/G purified from Bioreactor concentrate. | |
| Cell Surface | |
| THP-1 cells. Tonsil or Lymph Node |
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto