missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD115 Rat, APC, Clone: AFS98, enQuireBio™
Rat Monoclonal Antibody
Brand: EnquireBio QAB53-APC-100ug
This item is not returnable.
View return policy
Specifications
CD115 | |
Monoclonal | |
0.2mg/mL | |
10 mM NaH2PO4, 150 mM NaCl, 0.09% NaN3, 0.1% gelatin, pH7.2 | |
Csf1r | |
Protein A/G Purified | |
RUO | |
12978 | |
IgG2a κ |
Flow Cytometry | |
AFS98 | |
APC | |
XP_017173299 | |
Rat | |
100 μg | |
Primary | |
Store at 2-4°C. |