missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ CD 293 AGT™ Medium
CD 293 AGT™ is a protein-free, chemically-defined medium in an easy-to-use granular format.
108.00€ - 18140.00€
Specifications
Cell Line | 293 (HEK) |
---|---|
Culture Type | Suspension Cell Culture |
Form | Powder (AGT) |
Product Type | CD 293 Medium |
Serum Level | Serum-free |
Product Code | Brand | Quantity | Format | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Format | Price | Quantity & Availability | ||||
10145174
|
Gibco™
12529001 |
100 L | Ropak Bucket | N/A | |||||
10411995
|
Gibco™
12529003 |
10 kg | Ropak Bucket | N/A | |||||
11550436
|
Gibco™
12529012 |
10 L | Securitainer Capped Vial |
1026.00€
10 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10748224
|
Gibco™
12529020 |
1 L | Securitainer Capped Vial |
108.00€
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
CD 293 AGT™ is a protein-free, chemically-defined medium in an easy-to-use granular format. CD 293 AGT™ is optimized for the growth of suspension cultures of 293 cells.
AGT = Advanced Granulation Technology
Specifications
293 (HEK) | |
Powder (AGT) | |
Serum-free |
Suspension Cell Culture | |
CD 293 Medium | |
No antibiotics |
For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.