Learn More
CCS Viscosity Standard, ASTM D 5293 and SAE Specification J316, Paragon Scientific™
Paragon ISO 17025 / ISO 17034 certified CCS viscosity standards are for the calibration and verification of analytical equipment used in Cold Cranking Simulator (CCS) testing to ASTM D5293 and SAE Specification J300.
Brand: Paragon Scientific CL25
67.94 EUR valid until 2024-12-20
Use promo code "21695" to get your promotional price.
This item is not returnable.
View return policy
Description
- Certified according to ISO 17025 / ISO 17034 under UKAS accreditation
- Full Traceability to National Standards
- Ensures full compliance to ASTM & IP test method protocol
- Manufactured in the United Kingdom
- 2 year shelf life
Specifications
Glass Amber Bottle | |
Yes | |
Liquid |
Product Content Correction
Product Title
Nominal values quoted. Certified Values will vary from batch to batch.