Learn More
Novus Biologicals™ CCL15/MIP-1 delta Overexpression Lysate
Brand:  Novus Biologicals™ NBP2-05950
Product Code. 18078766
  • 685.73€ / 0.10mg
Explore more special offers
Description
Description
The CCL15/MIP-1 delta Overexpression Lysate is derived from human. It has been validated for the following applications: Western Blot.
Specifications
Specifications
The lysate was created in HEK293T cells, using plasmid ID RC215316 and based on accession number NP_11674. The protein contains a C-terminal DDK tag.
0.1 mg
Protein
-80°C. Avoid freeze-thaw cycles.
Western Blot
RIPA buffer
Homo sapiens Macrophage Inflammatory Protein 5, mRNA.
Native