Learn More
Novus Biologicals™ CCDC90B Overexpression Lysate
Brand:  Novus Biologicals™ NBP2-06515
Product Code. 18057049
  • 700.00€ / 0.10mg
Explore more special offers
Description
Description
The CCDC90B Overexpression Lysate is derived from human. It has been validated for the following applications: Western Blot.
Specifications
Specifications
The lysate was created in HEK293T cells, using plasmid ID RC215316 and based on accession number NP_68597. The protein contains a C-terminal DDK tag.
0.1 mg
Protein
-80°C. Avoid freeze-thaw cycles.
Native
Western Blot
RIPA buffer
Homo sapiens CCDC90B, mRNA.
metabolism